The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303484.1 | NP_001290413.1 | 8 - 9 | 1199 | 75 |
NM_031481.2 | NP_113669.1 | 9 - 10 | 1262 | 75 | |
XM_011546149.2 | XP_011544451.1 | 9 - 10 | 2191 | 75 | |
XM_011546150.2 | XP_011544452.1 | 9 - 10 | 2061 | 75 | |
XM_011546151.2 | XP_011544453.1 | 9 - 10 | 1450 | 75 | |
XM_011546152.1 | XP_011544454.1 | 9 - 10 | 1077 | 75 | |
XM_011546153.2 | XP_011544455.1 | 9 - 10 | 2072 | 75 | |
XM_011546154.2 | XP_011544456.1 | 9 - 10 | 1319 | 75 | |
XM_017028968.1 | XP_016884457.1 | 9 - 10 | 945 | 75 | |
XM_017028969.1 | XP_016884458.1 | 9 - 10 | 3091 | 75 | |
XM_017028970.1 | XP_016884459.1 | 10 - 11 | 2582 | 75 | |
XM_017028971.1 | XP_016884460.1 | 10 - 11 | 2571 | 75 | |
XM_017028972.1 | XP_016884461.1 | 10 - 11 | 2494 | 75 | |
XM_017028973.1 | XP_016884462.1 | 9 - 10 | 2519 | 75 | |
XM_017028974.1 | XP_016884463.1 | 10 - 11 | 2450 | 75 | |
XM_017028975.1 | XP_016884464.1 | 10 - 11 | 2362 | 75 | |
GenBank mRNA | AJ428203.1 | - | 9 - 10 | 1218 | 75 |
AY008285.1 | - | 9 - 10 | 1218 | 75 | |
BC031644.1 | - | 8 - 9 | 1154 | 75 | |
CR456578.1 | - | 7 - 8 | 848 | 75 |