Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPHLN1
Entrez Gene ID
51535
Gene Name
periphilin 1
Gene Aliases
CR, HSPC206, HSPC232
Chromosome Location
Chr.12: 42326126 - 42448621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143788.1 NP_001137260.1 2 - 3 179 72
NM_016488.6 NP_057572.5 2 - 3 179 72
NM_201439.1 NP_958847.1 2 - 3 179 72
NM_201515.1 NP_958923.1 3 - 4 222 72
XM_011538459.2 XP_011536761.1 3 - 4 200 72
XM_011538465.2 XP_011536767.1 2 - 3 157 72
XM_017019434.1 XP_016874923.1 3 - 4 200 72
XM_017019435.1 XP_016874924.1 3 - 4 199 72
XM_017019436.1 XP_016874925.1 3 - 4 200 72
XM_017019437.1 XP_016874926.1 2 - 3 157 72
XM_017019438.1 XP_016874927.1 2 - 3 157 72
XM_017019439.1 XP_016874928.1 3 - 4 200 72
XM_017019440.1 XP_016874929.1 3 - 4 200 72
XM_017019441.1 XP_016874930.1 2 - 3 157 72
XM_017019442.1 XP_016874931.1 2 - 3 157 72
XM_017019443.1 XP_016874932.1 3 - 4 200 72
XM_017019452.1 XP_016874941.1 3 - 4 200 72
GenBank mRNA AK055690.1 - 2 - 3 167 72
AK129910.1 - 3 - 4 222 72
AK308022.1 - 2 - 3 153 72
AY039238.1 - 2 - 3 161 72
AY157850.1 - 3 - 4 218 72

View Full Product Details