Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LTC4S
Entrez Gene ID
4056
Gene Name
leukotriene C4 synthase
Gene Aliases
-
Chromosome Location
Chr.5: 179793985 - 179796512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145867.1 NP_665874.1 3 - 4 65
GenBank mRNA AI480325.1 - 2 - 3 342 65
BC029498.1 - 3 - 4 65
BI597970.1 - 2 - 3 65
BI819419.1 - 2 - 3 65
U09353.1 - 3 - 4 65
U11552.1 - 3 - 4 65

View Full Product Details