The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002398.2 | NP_002389.1 | 2 - 3 | 697 | 111 |
XM_005264321.2 | XP_005264378.1 | 3 - 4 | 1020 | 111 | |
XM_005264322.2 | XP_005264379.1 | 2 - 3 | 972 | 111 | |
XM_005264323.2 | XP_005264380.1 | 3 - 4 | 1020 | 111 | |
XM_017004141.1 | XP_016859630.1 | 3 - 4 | 1020 | 111 | |
XM_017004142.1 | XP_016859631.1 | 2 - 3 | 972 | 111 | |
GenBank mRNA | AK297817.1 | - | 1 - 2 | 158 | 111 |
AK314949.1 | - | 2 - 3 | 447 | 111 | |
AL832770.1 | - | 2 - 3 | 710 | 111 | |
BC043503.1 | - | 2 - 3 | 692 | 111 | |
U85707.1 | - | 2 - 3 | 305 | 111 |