Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEIS1
Entrez Gene ID
4211
Gene Name
Meis homeobox 1
Gene Aliases
-
Chromosome Location
Chr.2: 66435125 - 66572765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002398.2 NP_002389.1 5 - 6 944 144
XM_005264321.2 XP_005264378.1 6 - 7 1267 144
XM_005264322.2 XP_005264379.1 5 - 6 1219 144
XM_005264323.2 XP_005264380.1 6 - 7 1267 144
XM_005264324.4 XP_005264381.1 4 - 5 340 144
XM_005264325.4 XP_005264382.1 4 - 5 1950 144
XM_017004141.1 XP_016859630.1 6 - 7 1267 144
XM_017004142.1 XP_016859631.1 5 - 6 1219 144
XM_017004143.1 XP_016859632.1 4 - 5 1950 144
GenBank mRNA AK297109.1 - 4 - 5 599 144
AK297817.1 - 4 - 5 405 144
AK314949.1 - 5 - 6 694 144
AL832770.1 - 5 - 6 957 144
BC043503.1 - 5 - 6 939 144
U85707.1 - 5 - 6 552 144

View Full Product Details