Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ADAR
- Entrez Gene ID
-
103
- Gene Name
- adenosine deaminase, RNA specific
- Gene Aliases
-
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
- Chromosome Location
-
Chr.1: 154582057 - 154628016 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_015841.3 |
NP_056656.2 |
5 - 6 |
2324 |
74 |
View Full Product Details