Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PATZ1
Entrez Gene ID
23598
Gene Name
POZ/BTB and AT hook containing zinc finger 1
Gene Aliases
MAZR, PATZ, RIAZ, ZBTB19, ZNF278, ZSG, dJ400N23
Chromosome Location
Chr.22: 31325804 - 31346263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032052.1 NP_114441.1 3 - 4 2164 91
GenBank mRNA AF254084.1 - 3 - 4 2133 91

View Full Product Details