The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111020.2 | NP_001104490.1 | 25 - 26 | 2666 | 74 |
NM_001130824.1 | NP_001124296.1 | 25 - 26 | 2591 | 74 | |
NM_001130825.1 | NP_001124297.1 | 24 - 25 | 2654 | 74 | |
NM_001319990.1 | NP_001306919.1 | 25 - 26 | 2588 | 74 | |
NM_001319991.1 | NP_001306920.1 | 24 - 25 | 2576 | 74 | |
NM_003169.3 | NP_003160.2 | 24 - 25 | 2730 | 74 | |
XM_017027174.1 | XP_016882663.1 | 20 - 21 | 2188 | 74 | |
GenBank mRNA | AB000516.1 | - | 25 - 26 | 2640 | 74 |
AB209257.1 | - | 23 - 24 | 2574 | 74 | |
AF040253.1 | - | 24 - 25 | 2694 | 74 | |
AF052165.1 | - | 4 - 5 | 447 | 74 | |
AK091476.1 | - | 12 - 13 | 2751 | 74 | |
AK296117.1 | - | 18 - 19 | 1928 | 74 | |
AK302954.1 | - | 26 - 27 | 2564 | 74 | |
AK303480.1 | - | 24 - 25 | 2445 | 74 | |
BC024203.2 | - | 25 - 26 | 2523 | 74 | |
Y12790.1 | - | 25 - 26 | 2630 | 74 |