Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICAM2
Entrez Gene ID
3384
Gene Name
intercellular adhesion molecule 2
Gene Aliases
CD102
Chromosome Location
Chr.17: 64002595 - 64020634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099789.1 NP_001093259.1 1 - 2 116 119
GenBank mRNA AB209878.1 - 1 - 2 224 119
AK295382.1 - 1 - 2 77 119
AK309953.1 - 1 - 2 114 119

View Full Product Details