The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 65 - 66 | 12552 | 87 |
NM_004662.2 | NP_004653.2 | 11 - 12 | 1949 | 87 | |
XM_011523703.1 | XP_011522005.1 | 65 - 66 | 12516 | 87 | |
XM_017024292.1 | XP_016879781.1 | 64 - 65 | 12399 | 87 | |
XM_017024293.1 | XP_016879782.1 | 54 - 55 | 10460 | 87 | |
GenBank mRNA | AB002355.2 | - | 45 - 46 | 8002 | 87 |
AB290158.1 | - | 52 - 53 | 10141 | 87 | |
AF015265.1 | - | 6 - 7 | 997 | 87 | |
AF257737.1 | - | 65 - 66 | 12542 | 87 | |
AJ404468.1 | - | 65 - 66 | 12552 | 87 | |
BC128261.1 | - | 9 - 10 | 1489 | 87 | |
BC128421.1 | - | 9 - 10 | 1489 | 87 | |
BC153883.1 | - | 45 - 46 | 8002 | 87 | |
X99947.1 | - | 11 - 12 | 1949 | 87 |