The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 25 - 26 | 5465 | 64 |
XM_011523703.1 | XP_011522005.1 | 25 - 26 | 5429 | 64 | |
XM_017024292.1 | XP_016879781.1 | 24 - 25 | 5312 | 64 | |
XM_017024293.1 | XP_016879782.1 | 14 - 15 | 3373 | 64 | |
XM_017024294.1 | XP_016879783.1 | 25 - 26 | 5429 | 64 | |
XM_017024295.1 | XP_016879784.1 | 25 - 26 | 5429 | 64 | |
GenBank mRNA | AB002355.2 | - | 6 - 7 | 1143 | 64 |
AB290158.1 | - | 13 - 14 | 3282 | 64 | |
AF257737.1 | - | 25 - 26 | 5455 | 64 | |
AJ404468.1 | - | 25 - 26 | 5465 | 64 | |
BC153883.1 | - | 6 - 7 | 1143 | 64 |