Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
Chromosome Location
Chr.17: 11598431 - 11970168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372.3 NP_001363.2 43 - 44 8491 113
XM_011523703.1 XP_011522005.1 43 - 44 8455 113
XM_017024292.1 XP_016879781.1 42 - 43 8338 113
XM_017024293.1 XP_016879782.1 32 - 33 6399 113
XM_017024294.1 XP_016879783.1 43 - 44 8455 113
GenBank mRNA AB002355.2 - 24 - 25 4169 113
AB290158.1 - 31 - 32 6308 113
AF257737.1 - 43 - 44 8481 113
AJ404468.1 - 43 - 44 8491 113
BC153883.1 - 24 - 25 4169 113

View Full Product Details