Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
Chromosome Location
Chr.17: 11598431 - 11970168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372.3 NP_001363.2 47 - 48 9085 83
XM_011523703.1 XP_011522005.1 47 - 48 9049 83
XM_017024292.1 XP_016879781.1 46 - 47 8932 83
XM_017024293.1 XP_016879782.1 36 - 37 6993 83
XM_017024294.1 XP_016879783.1 47 - 48 9049 83
GenBank mRNA AB002355.2 - 28 - 29 4763 83
AB290158.1 - 35 - 36 6902 83
AF257737.1 - 47 - 48 9075 83
AJ404468.1 - 47 - 48 9085 83
BC153883.1 - 28 - 29 4763 83

View Full Product Details