Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
Chromosome Location
Chr.17: 11598431 - 11970168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372.3 NP_001363.2 24 - 25 5220 70
XM_011523703.1 XP_011522005.1 24 - 25 5184 70
XM_017024292.1 XP_016879781.1 23 - 24 5067 70
XM_017024293.1 XP_016879782.1 13 - 14 3128 70
XM_017024294.1 XP_016879783.1 24 - 25 5184 70
XM_017024295.1 XP_016879784.1 24 - 25 5184 70
GenBank mRNA AB002355.2 - 5 - 6 898 70
AB290158.1 - 12 - 13 3037 70
AF257737.1 - 24 - 25 5210 70
AJ404468.1 - 24 - 25 5220 70
BC153883.1 - 5 - 6 898 70

View Full Product Details