The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015261.2 | NP_056076.1 | 5 - 6 | 1332 | 88 |
XM_011542719.2 | XP_011541021.1 | 4 - 5 | 593 | 88 | |
GenBank mRNA | AK124878.1 | - | 5 - 6 | 1332 | 88 |
BC098398.1 | - | 5 - 6 | 791 | 88 | |
BC144524.1 | - | 5 - 6 | 1016 | 88 | |
D29954.1 | - | 5 - 6 | 754 | 88 |