Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOCS1
Entrez Gene ID
4337
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
MIG11, MOCOD, MOCODA
Chromosome Location
Chr.6: 39904258 - 39934551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075098.3 NP_001068566.1 3 - 4 553 122
NM_005943.5 NP_005934.2 2 - 3 557 122
NR_033233.1 2 - 3 426 122
XM_011514632.2 XP_011512934.1 3 - 4 464 122
XM_011514633.2 XP_011512935.1 3 - 4 464 122
XM_011514634.2 XP_011512936.1 2 - 3 361 122
XM_017010888.1 XP_016866377.1 2 - 3 371 122
GenBank mRNA AF034374.1 - 3 - 4 436 122
AJ224328.1 - 2 - 3 554 122
AK056740.1 - 2 - 3 372 122
AK091306.1 - 2 - 3 285 122
AK300300.1 - 3 - 4 426 122
BC036839.1 - 3 - 4 426 122

View Full Product Details