The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018897.2 | NP_061720.2 | 4 - 5 | 349 | 149 |
XM_011511487.2 | XP_011509789.1 | 5 - 6 | 725 | 149 | |
XM_011511488.2 | XP_011509790.1 | 4 - 5 | 1519 | 149 | |
XM_011511489.2 | XP_011509791.1 | 3 - 4 | 311 | 149 | |
XM_011511490.2 | XP_011509792.1 | 4 - 5 | 1519 | 149 | |
XM_011511491.2 | XP_011509793.1 | 4 - 5 | 1521 | 149 | |
XM_011511492.2 | XP_011509794.1 | 4 - 5 | 1521 | 149 | |
XM_011511493.2 | XP_011509795.1 | 4 - 5 | 1521 | 149 | |
XM_011511494.2 | XP_011509796.1 | 4 - 5 | 1520 | 149 | |
XM_011511495.2 | XP_011509797.1 | 4 - 5 | 1520 | 149 | |
XM_017004504.1 | XP_016859993.1 | 4 - 5 | 1518 | 149 | |
GenBank mRNA | AB023161.2 | - | 4 - 5 | 367 | 149 |
AF327442.1 | - | 4 - 5 | 344 | 149 | |
AK094515.1 | - | 4 - 5 | 349 | 149 | |
BP199399.1 | - | 4 - 5 | 349 | 149 |