The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142386.2 | NP_001135858.1 | 3 - 4 | 543 | 98 |
NM_005391.4 | NP_005382.1 | 3 - 4 | 543 | 98 | |
GenBank mRNA | AK301965.1 | - | 3 - 4 | 546 | 98 |
BC015948.1 | - | 3 - 4 | 490 | 98 | |
BG717206.1 | - | 3 - 4 | 554 | 98 | |
L42452.1 | - | 3 - 4 | 336 | 98 |