Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD12
Entrez Gene ID
26090
Gene Name
abhydrolase domain containing 12
Gene Aliases
ABHD12A, BEM46L2, C20orf22, PHARC, dJ965G21.2
Chromosome Location
Chr.20: 25294743 - 25390982 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042472.2 NP_001035937.1 6 - 7 895 70
NM_015600.4 NP_056415.1 6 - 7 895 70
XM_005260698.1 XP_005260755.1 6 - 7 626 70
XM_011529214.2 XP_011527516.1 6 - 7 626 70
XM_011529215.1 XP_011527517.1 7 - 8 746 70
XM_011529218.1 XP_011527520.1 4 - 5 305 70
XM_017027796.1 XP_016883285.1 7 - 8 746 70
XM_017027798.1 XP_016883287.1 5 - 6 425 70
GenBank mRNA AK075023.1 - 6 - 7 626 70
AK290815.1 - 6 - 7 626 70
AK293495.1 - 7 - 8 512 70
AL117442.1 - 3 - 4 163 70
BC014049.2 - 6 - 7 740 70

View Full Product Details