Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYN1
Entrez Gene ID
6853
Gene Name
synapsin I
Gene Aliases
SYN1a, SYN1b, SYNI
Chromosome Location
Chr.X: 47571901 - 47619857 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006950.3 NP_008881.2 10 - 11 1434 120
NM_133499.2 NP_598006.1 10 - 11 1434 120
GenBank mRNA AL833961.1 - 2 - 3 257 120

View Full Product Details