The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133499.2 | NP_598006.1 | 12 - 13 | 2111 | 122 |
GenBank mRNA | AL833961.1 | - | 4 - 5 | 934 | 122 |