The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007058.3 | NP_008989.2 | 19 - 20 | 1972 | 96 |
XM_006714984.1 | XP_006715047.1 | 19 - 20 | 1975 | 96 | |
XM_006714985.1 | XP_006715048.1 | 19 - 20 | 1975 | 96 | |
XM_011514274.1 | XP_011512576.1 | 19 - 20 | 2056 | 96 | |
XM_011514275.1 | XP_011512577.1 | 20 - 21 | 2098 | 96 | |
GenBank mRNA | AJ242832.1 | - | 18 - 19 | 1926 | 96 |
AK314054.1 | - | 19 - 20 | 1952 | 96 | |
BC033733.1 | - | 19 - 20 | 1943 | 96 |