The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001217.4 | NP_001208.2 | 4 - 5 | 1156 | 60 |
NR_136241.1 | 4 - 5 | 1156 | 60 | ||
GenBank mRNA | AB018195.1 | - | 4 - 5 | 845 | 60 |
AF050106.1 | - | 2 - 3 | 335 | 60 | |
AF067662.1 | - | 4 - 5 | 840 | 60 | |
AY358967.1 | - | 4 - 5 | 1061 | 60 | |
BC002662.2 | - | 4 - 5 | 571 | 60 | |
BT007265.1 | - | 4 - 5 | 476 | 60 | |
CR541772.1 | - | 4 - 5 | 476 | 60 |