The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001217.4 | NP_001208.2 | 5 - 6 | 1248 | 74 |
NR_136241.1 | 5 - 6 | 1248 | 74 | ||
GenBank mRNA | AF067662.1 | - | 5 - 6 | 932 | 74 |
AY358967.1 | - | 5 - 6 | 1153 | 74 | |
BC002662.2 | - | 5 - 6 | 663 | 74 | |
BT007265.1 | - | 5 - 6 | 568 | 74 | |
CR541772.1 | - | 5 - 6 | 568 | 74 |