Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CA11
Entrez Gene ID
770
Gene Name
carbonic anhydrase 11
Gene Aliases
CARPX1
Chromosome Location
Chr.19: 48637942 - 48646312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001217.4 NP_001208.2 6 - 7 1327 62
NR_136241.1 6 - 7 1327 62
GenBank mRNA AB018195.1 - 6 - 7 1016 62
AF050106.1 - 4 - 5 506 62
AF067662.1 - 6 - 7 1011 62
AY358967.1 - 6 - 7 1232 62
BC002662.2 - 6 - 7 742 62
BT007265.1 - 6 - 7 647 62
CR541772.1 - 6 - 7 647 62

View Full Product Details