Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA11
Entrez Gene ID
770
Gene Name
carbonic anhydrase 11
Gene Aliases
CARPX1
Chromosome Location
Chr.19: 48637942 - 48646312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001217.4 NP_001208.2 8 - 9 1639 79
GenBank mRNA AB018195.1 - 8 - 9 1328 79
AI568635.1 - 1 - 2 79
AY358967.1 - 8 - 9 1544 79
BC002662.2 - 8 - 9 1054 79
HY313058.1 - 1 - 2 79

View Full Product Details