The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042600.2 | NP_001036065.1 | 8 - 9 | 676 | 61 |
NM_007181.5 | NP_009112.1 | 8 - 9 | 676 | 61 | |
XM_011526403.1 | XP_011524705.1 | 8 - 9 | 574 | 61 | |
XM_011526404.1 | XP_011524706.1 | 8 - 9 | 574 | 61 | |
XM_017026231.1 | XP_016881720.1 | 8 - 9 | 582 | 61 | |
GenBank mRNA | AK303267.1 | - | 1 - 2 | 51 | 61 |
BX345201.2 | - | 8 - 9 | 644 | 61 | |
U66464.1 | - | 8 - 9 | 590 | 61 |