Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
Chromosome Location
Chr.7: 2537810 - 2555758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011515184.2 XP_011513486.1 1 - 2 112 66

View Full Product Details