Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
Chromosome Location
Chr.7: 2537810 - 2555758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152743.3 NP_689956.2 4 - 5 721 56
XM_005249643.4 XP_005249700.1 4 - 5 683 56
XM_011515177.2 XP_011513479.1 4 - 5 688 56
XM_011515178.1 XP_011513480.1 4 - 5 899 56
XM_011515179.2 XP_011513481.1 4 - 5 679 56
XM_011515181.2 XP_011513483.1 4 - 5 679 56
XM_011515182.2 XP_011513484.1 4 - 5 474 56
XM_011515184.2 XP_011513486.1 4 - 5 474 56
XM_011515186.2 XP_011513488.1 4 - 5 680 56
XM_017011833.1 XP_016867322.1 4 - 5 484 56
XM_017011834.1 XP_016867323.1 4 - 5 675 56
XM_017011836.1 XP_016867325.1 4 - 5 677 56
GenBank mRNA AK024482.1 - 4 - 5 469 56
AK131083.1 - 4 - 5 657 56
BC015632.2 - 4 - 5 475 56

View Full Product Details