The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152743.3 | NP_689956.2 | 6 - 7 | 1217 | 109 |
XM_005249643.4 | XP_005249700.1 | 6 - 7 | 1179 | 109 | |
XM_011515177.2 | XP_011513479.1 | 6 - 7 | 1184 | 109 | |
XM_011515178.1 | XP_011513480.1 | 6 - 7 | 1395 | 109 | |
XM_011515179.2 | XP_011513481.1 | 6 - 7 | 1175 | 109 | |
XM_011515181.2 | XP_011513483.1 | 6 - 7 | 1175 | 109 | |
XM_011515182.2 | XP_011513484.1 | 6 - 7 | 970 | 109 | |
XM_011515183.1 | XP_011513485.1 | 5 - 6 | 1064 | 109 | |
XM_011515184.2 | XP_011513486.1 | 6 - 7 | 970 | 109 | |
XM_011515186.2 | XP_011513488.1 | 6 - 7 | 1176 | 109 | |
XM_017011833.1 | XP_016867322.1 | 6 - 7 | 980 | 109 | |
XM_017011834.1 | XP_016867323.1 | 6 - 7 | 1171 | 109 | |
XM_017011835.1 | XP_016867324.1 | 5 - 6 | 1064 | 109 | |
XM_017011836.1 | XP_016867325.1 | 6 - 7 | 1173 | 109 | |
GenBank mRNA | AK024482.1 | - | 5 - 6 | 1231 | 109 |
BC007209.1 | - | 2 - 3 | 452 | 109 | |
BC015632.2 | - | 6 - 7 | 971 | 109 | |
BC023561.2 | - | 2 - 3 | 212 | 109 |