Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLG
Entrez Gene ID
5428
Gene Name
DNA polymerase gamma, catalytic subunit
Gene Aliases
MDP1, MIRAS, MTDPS4A, MTDPS4B, PEO, POLG1, POLGA, SANDO, SCAE
Chromosome Location
Chr.15: 89316305 - 89334795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126131.1 NP_001119603.1 13 - 14 2536 85
NM_002693.2 NP_002684.1 13 - 14 2548 85
GenBank mRNA AK291281.1 - 13 - 14 2567 85
BC042571.1 - 13 - 14 2533 85
BC050559.1 - 13 - 14 2517 85
D84103.1 - 12 - 13 2372 85
U60325.1 - 12 - 13 2425 85
X98093.1 - 13 - 14 2548 85

View Full Product Details