The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126131.1 | NP_001119603.1 | 14 - 15 | 2701 | 123 |
NM_002693.2 | NP_002684.1 | 14 - 15 | 2713 | 123 | |
GenBank mRNA | AK291281.1 | - | 14 - 15 | 2732 | 123 |
AL110092.1 | - | 2 - 3 | 192 | 123 | |
BC042571.1 | - | 14 - 15 | 2698 | 123 | |
BC050559.1 | - | 14 - 15 | 2682 | 123 | |
D84103.1 | - | 13 - 14 | 2537 | 123 | |
U60325.1 | - | 13 - 14 | 2590 | 123 | |
X98093.1 | - | 14 - 15 | 2713 | 123 |