Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLG
Entrez Gene ID
5428
Gene Name
DNA polymerase gamma, catalytic subunit
Gene Aliases
MDP1, MIRAS, MTDPS4A, MTDPS4B, PEO, POLG1, POLGA, SANDO, SCAE
Chromosome Location
Chr.15: 89316305 - 89334795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126131.1 NP_001119603.1 21 - 22 3749 67
NM_002693.2 NP_002684.1 21 - 22 3761 67
GenBank mRNA AK130624.1 - 4 - 5 635 67
AK291281.1 - 21 - 22 3780 67
BC042571.1 - 21 - 22 3746 67
BC050559.1 - 21 - 22 3730 67
D84103.1 - 20 - 21 3585 67
U60325.1 - 20 - 21 3638 67
X98093.1 - 21 - 22 3761 67

View Full Product Details