The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126131.1 | NP_001119603.1 | 8 - 9 | 1853 | 94 |
NM_002693.2 | NP_002684.1 | 8 - 9 | 1865 | 94 | |
GenBank mRNA | AK291281.1 | - | 8 - 9 | 1884 | 94 |
BC042571.1 | - | 8 - 9 | 1850 | 94 | |
BC050559.1 | - | 8 - 9 | 1834 | 94 | |
D84103.1 | - | 7 - 8 | 1689 | 94 | |
U60325.1 | - | 7 - 8 | 1742 | 94 | |
X98093.1 | - | 8 - 9 | 1865 | 94 |