Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERPINF2
Entrez Gene ID
5345
Gene Name
serpin family F member 2
Gene Aliases
A2AP, AAP, ALPHA-2-PI, API, PLI
Chromosome Location
Chr.17: 1742808 - 1755265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000934.3 NP_000925.2 4 - 5 239 141
NM_001165920.1 NP_001159392.1 4 - 5 217 141
XM_005256701.4 XP_005256758.2 4 - 5 311 141
XM_017024765.1 XP_016880254.1 5 - 6 326 141
GenBank mRNA BC031592.1 - 4 - 5 217 141
BX107460.1 - 4 - 5 239 141
D00174.1 - 3 - 4 186 141
J02654.1 - 3 - 4 153 141

View Full Product Details