Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC43A2
Entrez Gene ID
124935
Gene Name
solute carrier family 43 member 2
Gene Aliases
LAT4
Chromosome Location
Chr.17: 1569254 - 1630014 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284498.1 NP_001271427.1 4 - 5 762 75
NM_001321364.1 NP_001308293.1 4 - 5 560 75
NM_001321365.1 NP_001308294.1 4 - 5 729 75
NM_152346.2 NP_689559.1 4 - 5 560 75
XM_017024177.1 XP_016879666.1 4 - 5 737 75
XM_017024178.1 XP_016879667.1 4 - 5 770 75
XM_017024179.1 XP_016879668.1 3 - 4 669 75
GenBank mRNA AB120364.1 - 4 - 5 728 75
AF289592.1 - 4 - 5 605 75
AK075068.1 - 2 - 3 100 75
AK075173.1 - 2 - 3 154 75
BC027923.1 - 4 - 5 510 75
BC071859.1 - 4 - 5 560 75

View Full Product Details