The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004959.4 | NP_004950.2 | 3 - 4 | 430 | 75 |
XM_005251871.3 | XP_005251928.1 | 3 - 4 | 458 | 75 | |
XM_011518455.2 | XP_011516757.1 | 3 - 4 | 308 | 75 | |
GenBank mRNA | AB307718.1 | - | 2 - 3 | 243 | 75 |
BC032501.1 | - | 3 - 4 | 430 | 75 | |
HQ692856.1 | - | 2 - 3 | 243 | 75 | |
HQ709184.1 | - | 2 - 3 | 243 | 75 | |
U76388.1 | - | 2 - 3 | 258 | 75 |