The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013318.3 | NP_037450.2 | 8 - 9 | 1174 | 91 |
GenBank mRNA | AK309879.1 | - | 1 - 2 | 123 | 91 |
CR749818.1 | - | 2 - 3 | 249 | 91 |