The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316345.1 | NP_001303274.1 | 18 - 19 | 2288 | 103 |
NM_021111.2 | NP_066934.1 | 16 - 17 | 2152 | 103 | |
XM_017015207.1 | XP_016870696.1 | 17 - 18 | 2188 | 103 | |
XM_017015208.1 | XP_016870697.1 | 17 - 18 | 2262 | 103 | |
XM_017015209.1 | XP_016870698.1 | 17 - 18 | 2251 | 103 | |
XM_017015210.1 | XP_016870699.1 | 15 - 16 | 2003 | 103 | |
GenBank mRNA | AK292653.1 | - | 16 - 17 | 2152 | 103 |
AK315073.1 | - | 16 - 17 | 2138 | 103 | |
BC137093.1 | - | 16 - 17 | 2113 | 103 | |
D50406.1 | - | 16 - 17 | 2175 | 103 |