Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RECK
Entrez Gene ID
8434
Gene Name
reversion inducing cysteine rich protein with kazal motifs
Gene Aliases
ST15
Chromosome Location
Chr.9: 36036905 - 36124455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316346.1 NP_001303275.1 6 - 7 492 97
NM_001316347.1 NP_001303276.1 6 - 7 492 97
NM_001316348.1 NP_001303277.1 6 - 7 492 97
NM_021111.2 NP_066934.1 6 - 7 492 97
XM_017015207.1 XP_016870696.1 7 - 8 528 97
XM_017015211.1 XP_016870700.1 6 - 7 494 97
GenBank mRNA AK292653.1 - 6 - 7 492 97
AK315073.1 - 6 - 7 478 97
BC060806.1 - 6 - 7 466 97
BC137093.1 - 6 - 7 453 97
D50406.1 - 6 - 7 515 97
JQ756320.1 - 6 - 7 406 97

View Full Product Details