The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031468.3 | NP_113656.2 | 2 - 3 | 510 | 123 |
XM_017012675.1 | XP_016868164.1 | 3 - 4 | 865 | 123 | |
XM_017012676.1 | XP_016868165.1 | 2 - 3 | 647 | 123 | |
GenBank mRNA | AY007302.1 | - | 1 - 2 | 168 | 123 |
BG718538.1 | - | 2 - 3 | 515 | 123 |