The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000663.4 | NP_000654.2 | 5 - 6 | 1257 | 91 |
NM_001127448.1 | NP_001120920.1 | 5 - 6 | 579 | 91 | |
NM_020686.5 | NP_065737.2 | 5 - 6 | 485 | 91 | |
XM_011522400.2 | XP_011520702.1 | 6 - 7 | 825 | 91 | |
XM_011522401.2 | XP_011520703.1 | 6 - 7 | 786 | 91 | |
GenBank mRNA | AK290501.1 | - | 5 - 6 | 91 | |
AK293983.1 | - | 5 - 6 | 626 | 91 | |
AK310771.1 | - | 5 - 6 | 91 | ||
BC015628.2 | - | 5 - 6 | 406 | 91 | |
BC031413.1 | - | 5 - 6 | 437 | 91 | |
BG749438.1 | - | 5 - 6 | 407 | 91 | |
BI459888.1 | - | 5 - 6 | 490 | 90 | |
BX537362.1 | - | 5 - 6 | 91 | ||
KJ534758.1 | - | 4 - 5 | 319 | 91 | |
KJ535005.1 | - | 4 - 5 | 319 | 91 | |
S75578.1 | - | 3 - 4 | 214 | 91 | |
U80226.1 | - | 4 - 5 | 91 |