Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MYO15A
- Entrez Gene ID
-
51168
- Gene Name
- myosin XVA
- Gene Aliases
-
DFNB3, MYO15
- Chromosome Location
-
Chr.17: 18108706 - 18180227 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK055873.1 |
- |
1 - 1 |
1880 |
136 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD512374.1 |
- |
|
119 |
|
Chr. - - [-] on Build |
136 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
GID complex subunit 4 homolog |
GID4 |
C17orf39, VID24 |
-38.9kb
|
Anti-sense |
Chr. - 18039297 - 18068405 on Build GRCh38 |
Downstream |
alkB homolog 5, RNA demethylase |
ALKBH5 |
ABH5, OFOXD, OFOXD1 |
73.1kb
|
Anti-sense |
Chr. - 18183553 - 18209954 on Build GRCh38 |
View Full Product Details