The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002609.3 | NP_002600.1 | 8 - 9 | 1717 | 71 |
XM_005268464.2 | XP_005268521.1 | 7 - 8 | 1696 | 71 | |
XM_011537658.1 | XP_011535960.1 | 8 - 9 | 1292 | 71 | |
XM_011537659.1 | XP_011535961.1 | 9 - 10 | 2197 | 71 | |
GenBank mRNA | AB209657.1 | - | 8 - 9 | 1750 | 71 |
AK293093.1 | - | 8 - 9 | 1553 | 71 | |
BC032224.1 | - | 8 - 9 | 1723 | 71 | |
J03278.1 | - | 8 - 9 | 1604 | 71 | |
M21616.1 | - | 8 - 9 | 1434 | 71 |