Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
Chromosome Location
Chr.17: 4163371 - 4264026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016376.3 NP_057460.3 1 - 2 52 66
NR_047571.1 1 - 2 52 66
XM_005256679.3 XP_005256736.1 1 - 2 99 66
GenBank mRNA AK025483.1 - 1 - 2 29 66
AK057047.1 - 1 - 2 29 66
AK225170.1 - 1 - 2 29 66
AK292930.1 - 1 - 2 29 66
AK302712.1 - 1 - 2 38 66
AL706226.1 - 1 - 2 53 66
DA856905.1 - 1 - 2 52 66

View Full Product Details