Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
Chromosome Location
Chr.17: 4163371 - 4264026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257999.1 NP_001244928.1 9 - 10 1410 76
NM_016376.3 NP_057460.3 9 - 10 1205 76
NR_047571.1 8 - 9 1086 76
XM_005256679.3 XP_005256736.1 9 - 10 1252 76
XM_011523926.1 XP_011522228.1 9 - 10 1308 76
XM_011523928.2 XP_011522230.1 9 - 10 1290 76
XM_011523933.1 XP_011522235.1 5 - 6 694 76
XM_017024733.1 XP_016880222.1 9 - 10 1505 76
GenBank mRNA AB033081.2 - 9 - 10 1410 76
AK025483.1 - 9 - 10 1182 76
AK057047.1 - 8 - 9 1063 76
AK225170.1 - 9 - 10 1182 76
AK292930.1 - 9 - 10 1182 76
BC052308.1 - 9 - 10 1178 76
CR933717.1 - 3 - 4 732 76

View Full Product Details