The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004140.3 | NP_004131.3 | 14 - 15 | 1964 | 68 |
XM_011523849.2 | XP_011522151.1 | 14 - 15 | 1993 | 68 | |
XM_011523850.2 | XP_011522152.1 | 14 - 15 | 1967 | 68 | |
XM_011523851.2 | XP_011522153.1 | 14 - 15 | 1993 | 68 | |
XM_011523852.2 | XP_011522154.1 | 14 - 15 | 1993 | 68 | |
XM_011523853.2 | XP_011522155.1 | 14 - 15 | 1993 | 68 | |
XM_011523854.2 | XP_011522156.1 | 14 - 15 | 1993 | 68 | |
XM_011523855.2 | XP_011522157.1 | 14 - 15 | 1993 | 68 | |
XM_011523856.1 | XP_011522158.1 | 14 - 15 | 1993 | 68 | |
GenBank mRNA | BC151838.1 | - | 14 - 15 | 1940 | 68 |
BQ937224.1 | - | 5 - 6 | 666 | 68 | |
D50550.1 | - | 14 - 15 | 1948 | 68 |