Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEIS1
Entrez Gene ID
4211
Gene Name
Meis homeobox 1
Gene Aliases
-
Chromosome Location
Chr.2: 66435125 - 66572765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005264321.2 XP_005264378.1 12 - 13 1891 64
XM_005264322.2 XP_005264379.1 11 - 12 1843 64
XM_005264324.4 XP_005264381.1 10 - 11 964 64
XM_017004144.1 XP_016859633.1 11 - 12 1749 64
XM_017004145.1 XP_016859634.1 10 - 11 1701 64
GenBank mRNA AK297817.1 - 9 - 10 883 64

View Full Product Details