The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017602.3 | NP_060072.1 | 4 - 5 | 982 | 83 |
XM_005278039.3 | XP_005278096.1 | 4 - 5 | 1732 | 83 | |
XM_005278042.4 | XP_005278099.1 | 4 - 5 | 477 | 83 | |
XM_006724537.2 | XP_006724600.1 | 4 - 5 | 1732 | 83 | |
XM_011543932.1 | XP_011542234.1 | 4 - 5 | 1732 | 83 | |
XM_011543934.2 | XP_011542236.1 | 4 - 5 | 495 | 83 | |
XM_017029629.1 | XP_016885118.1 | 5 - 6 | 476 | 83 | |
XM_017029630.1 | XP_016885119.1 | 4 - 5 | 477 | 83 | |
GenBank mRNA | AK022612.1 | - | 1 - 2 | 165 | 83 |
BC009917.2 | - | 4 - 5 | 982 | 83 |