The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286124.1 | NP_001273053.1 | 3 - 4 | 940 | 220 |
NM_001286126.1 | NP_001273055.1 | 3 - 4 | 940 | 220 | |
NM_001286128.1 | NP_001273057.1 | 3 - 4 | 940 | 154 | |
NM_018650.4 | NP_061120.3 | 3 - 4 | 940 | 220 | |
XM_005273134.4 | XP_005273191.1 | 3 - 4 | 807 | 220 | |
XM_011509561.2 | XP_011507863.1 | 3 - 4 | 936 | 220 | |
XM_017001305.1 | XP_016856794.1 | 4 - 5 | 397 | 220 | |
XM_017001307.1 | XP_016856796.1 | 4 - 5 | 837 | 220 | |
GenBank mRNA | AB040910.1 | - | 3 - 4 | 897 | 154 |
AF154845.1 | - | 3 - 4 | 566 | 220 | |
AK295506.1 | - | 3 - 4 | 438 | 220 | |
AK314614.1 | - | 3 - 4 | 600 | 220 | |
BC113869.1 | - | 3 - 4 | 476 | 220 | |
BC114478.1 | - | 3 - 4 | 476 | 220 | |
KJ534879.1 | - | 3 - 4 | 300 | 220 | |
KJ534881.1 | - | 3 - 4 | 300 | 220 | |
KJ535089.1 | - | 3 - 4 | 299 | 220 |